2-Phenyl-5-benzimidazolesulfonic acid

Supplier: APOLLO SCIENTIFIC
Danger

OR28712-25G OR28712-5G
77408-178EA 36.68 USD
77408-178 77408-180
2-Phenyl-5-benzimidazolesulfonic acid
2-Phenyl-5-benzimidazolesulfonic acid
Formula: C₁₃H₁₀N₂O₃S
Storage Temperature: Ambient
MDL Number: MFCD00053007
CAS Number: 27503-81-7
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR