2-Chloro-5-hydroxybenzoic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR2647-1G OR2647-5G
77407-790EA 96.78 USD
77407-790 77407-792
2-Chloro-5-hydroxybenzoic acid 98%
2-Chloro-5-hydroxybenzoic acid
Formula: C₇H₅ClO₃
Melting Pt: 178…180 °C
Storage Temperature: Ambient
MDL Number: MFCD04038818
CAS Number: 56961-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR