1-Bromo-5-chloropentane

Supplier: APOLLO SCIENTIFIC
Warning

OR24918-100G OR24918-25G
77407-558EA 23.36 USD
77407-558 77407-560
1-Bromo-5-chloropentane
1-Bromo-5-chloropentane
Formula: BrH₂C(CH₂)₃CH₂Cl
Boiling Pt: 210…212 °C
Density: 1.408 g/cm³ (25 °C)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00001016
CAS Number: 54512-75-3
EINECS: 259-193-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR