N,N'-Dimethylethylendiamine 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: N1,N2-Dimethylethane-1,2-diamine, sym. Dimethylethylendiamine, N,N'-Dimethylethylenediamine

OR322130-100G OR322130-25G OR322130-5G
77409-154EA 108 USD
77409-154 77409-156 77409-158
N,N'-Dimethylethylendiamine 95%
N,N'-Dimethylethylendiamine
Formula: C₄H₁₂N₂
MW: 88.15 g/mol
Boiling Pt: 118…119 °C
Density: 0.818 g/cm³ (25 °C)
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00008290
CAS Number: 110-70-3
EINECS: 230-793-3
UN: 2734
ADR: 8+3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR