7-Methoxy-3,4,5,6-tetrahydro-2H-azepine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Aza-2-methoxy-1-cycloheptene, O-Methylcaprolactim, 7-Methoxy-3,4,5,6-tetrahydro-2H-azepine

OR30218-25G OR30218-5G
77408-828EA 265.92 USD
77408-828 77408-830
7-Methoxy-3,4,5,6-tetrahydro-2H-azepine
7-Methoxy-3,4,5,6-tetrahydro-2H-azepine
Formula: C₇H₁₃NO
MW: 127.19 g/mol
Boiling Pt: 52…53 °C/18 mmHg
Density: 0.887 g/cm³ (25 °C)
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00006938
CAS Number: 2525-16-8
EINECS: 219-762-2
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR