(S)-(+)-2-Amino-1-phenylethanol 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR3013-5G OR3013-1G OR3013-25G
77408-814EA 104.94 USD
77408-814 77408-810 77408-812
(S)-(+)-2-Amino-1-phenylethanol 98%
(S)-(+)-2-Amino-1-phenylethanol
Formula: C₈H₁₁NO
MW: 137.182 g/mol
Boiling Pt: 160 °C/17 mmHg
Melting Pt: 57…63 °C
Storage Temperature: Freezer
MDL Number: MFCD00239405
CAS Number: 56613-81-1
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR