2,6-Diaminopyrimidin-4(3H)-one 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,4-Diamino-1,6-dihydro-6-oxopyrimidine

OR26562-1G OR26562-250MG
77407-808EA 30 USD
77407-808 77407-810 77407-814
2,6-Diaminopyrimidin-4(3H)-one 98%
2,6-Diaminopyrimidin-4(3H)-one
Formula: C₄H₆N₄O
MW: 126.117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00043013
CAS Number: 143504-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR