2-Bromo-4-methylthiazole

Supplier: APOLLO SCIENTIFIC
Warning

OR2600-1G OR2600-5G
77407-724EA 23.2 USD
77407-724 77407-726
2-Bromo-4-methylthiazole
2-Bromo-4-methylthiazole
Formula: C₄H₄BrNS
Storage Temperature: Ambient
MDL Number: MFCD06660130
CAS Number: 7238-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR