5-(4-Methoxyphenyl)-1,3-oxazole-4-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR25969-250MG OR25969-100MG
77407-714EA 186.43 USD
77407-714 77407-716 77407-712
5-(4-Methoxyphenyl)-1,3-oxazole-4-carboxylic acid
5-(4-Methoxyphenyl)-1,3-oxazole-4-carboxylic acid
Formula: C₁₁H₉NO₄
Melting Pt: 147…151 °C
Storage Temperature: Ambient
MDL Number: MFCD03644147
CAS Number: 89205-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR