3-Methylenecyclobutanecarbonitrile

Supplier: APOLLO SCIENTIFIC
Warning

OR2438-25G OR2438-5G
77407-480EA 141.62 USD
77407-480 77407-482
3-Methylenecyclobutanecarbonitrile
3-Methylenecyclobutanecarbonitrile
Formula: C₆H₇N
MW: 93.13 g/mol
Boiling Pt: 149…150 °C
Storage Temperature: Ambient
MDL Number: MFCD01085401
CAS Number: 15760-35-7
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR