4-Acetoxyindole 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
4-Indolyl acetate
OR2401-100G
OR2401-25G
OR2401-5G
77407-398EA
690.76
USD
77407-398
77407-404
77407-406
4-Acetoxyindole 98%
4-Acetoxyindole
Formula:
C₁₀H₉NO₂ MW: 175.19 g/mol Melting Pt: 98…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010678 CAS Number: 5585-96-6 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...