2-Aminofuran, N-BOC protected 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-[(tert-Butoxycarbonyl)amino]furan, 2-(Boc-amino)furan, tert-Butyl (fur-2-yl)carbamate

OR22968-1G OR22968-250MG
77406-984EA 58.08 USD
77406-984 77406-986
2-Aminofuran, N-BOC protected 97%
2-Aminofuran, N-BOC protected
Formula: C₉H₁₃NO₃
MW: 183.204 g/mol
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD02682023
CAS Number: 56267-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR