4-Bromo-2,6-dichloroaniline 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR21224-100G OR21224-25G
77406-650EA 80.8 USD
77406-650 77406-652
4-Bromo-2,6-dichloroaniline 98%
4-Bromo-2,6-dichloroaniline
Formula: C₆H₄BrCl₂N
Melting Pt: 85…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00025162
CAS Number: 697-88-1
EINECS: 211-809-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR