3-[(Pyrrolidin-1-yl)methyl]benzeneboronic acid, pinacol ester 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)pyrrolidine, 1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]pyrrolidine, 3-(Pyrrolidinomethyl)phenylboronic acid, pinacol ester

OR2072-1G OR2072-250MG
77406-566EA 131.42 USD
77406-566 77406-568
3-[(Pyrrolidin-1-yl)methyl]benzeneboronic acid, pinacol ester 97%
3-[(Pyrrolidin-1-yl)methyl]benzeneboronic acid, pinacol ester
Formula: C₁₇H₂₆BNO₂
MW: 287.205 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08741425
CAS Number: 884507-45-3
UN: 2922
ADR: 8+6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR