7-Bromothieno[3,2-b]pyridine

Supplier: APOLLO SCIENTIFIC
Warning

OR300381-1G OR300381-250MG
77408-616EA 353.86 USD
77408-616 77408-618
7-Bromothieno[3,2-b]pyridine
7-Bromothieno[3,2-b]pyridine
Formula: C₇H₄BrNS
MW: 214.082 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18801029
CAS Number: 603305-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR