Pyrrolidine-1-carbonitrile 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-Cyanopyrrolidine

OR28605-100MG OR28605-25G OR28605-5G
77408-108EA 21.26 USD
77408-108 77408-110 77408-112
Pyrrolidine-1-carbonitrile 97%
Pyrrolidine-1-carbonitrile
Formula: C₅H₈N₂
MW: 96.13 g/mol
Boiling Pt: 75…77 °C/1.8 mmHg
Density: 0.954 g/cm³ (25 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00003165
CAS Number: 1530-88-7
EINECS: 216-234-3
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR