2-Isopropylaniline 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR28577-100G
OR28577-500G
OR28577-5G
77408-100EA
108
USD
77408-100
77408-102
77408-104
2-Isopropylaniline 97%
2-Isopropylaniline
Formula:
C₉H₁₃N Boiling Pt: 214…216 °C/10 mmHg Density: 0.966 g/cm³ (25 °C) Flash Pt: 95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007720 CAS Number: 643-28-7 EINECS: 211-397-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...