2-Methylthiophene-3-carboxaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Methylthiophene-3-carbaldehyde, 3-Formyl-2-methylthiophene

OR2624-1G OR2624-250MG
77407-766EA 737.62 USD
77407-766 77407-768
2-Methylthiophene-3-carboxaldehyde
2-Methylthiophene-3-carboxaldehyde
Formula: C₆H₆OS
Boiling Pt: 80.5…82 °C (10 mmHg)
Storage Temperature: Refrigerator
MDL Number: MFCD09864856
CAS Number: 84815-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR