4-Iodopyridine 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR25896-1G OR25896-5G
77407-682EA 21.32 USD
77407-682 77407-684
4-Iodopyridine 97%
4-Iodopyridine
Formula: C₅H₄IN
MW: 205 g/mol
Melting Pt: 92…96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093937
CAS Number: 15854-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR