2,5-Dichloro-4-pyridineboronic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR2585-1G OR2585-250MG OR2585-5G
77407-674EA 49.92 USD
77407-674 77407-676 77407-678
2,5-Dichloro-4-pyridineboronic acid
2,5-Dichloro-4-pyridineboronic acid
Formula: C₅H₄BCl₂NO₂
Melting Pt: 161…163 °C
Storage Temperature: Ambient
MDL Number: MFCD06798254
CAS Number: 847664-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR