3-Oxo-3-(2-thienyl)propionitrile

Supplier: APOLLO SCIENTIFIC
Warning

OR23646-5G OR23646-10G OR23646-1G
77407-354EA 62.8 USD
77407-354 77407-350 77407-352
3-Oxo-3-(2-thienyl)propionitrile
3-Oxo-3-(2-thienyl)propionitrile
Formula: C₇H₅NOS
Storage Temperature: Ambient
MDL Number: MFCD00052586
CAS Number: 33898-90-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR