tert-Butyl nitrite 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR2337-100G OR2337-25G OR2337-500G
77407-288EA 67.26 USD
77407-288 77407-292 77407-294
tert-Butyl nitrite 95%
tert-Butyl nitrite
Formula: C₄H₉NO₂
Boiling Pt: 61…63 °C
Density: 0.867 g/cm³ (25 °C)
Flash Pt: –10 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002055
CAS Number: 540-80-7
EINECS: 208-757-0
UN: 2351
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR