1,3-dimethyl-1H-thieno[2,3-c]pyrazole-5-carboxylic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1,3-Dimethyl-1H-thieno[2,3-c]pyrazole-5-carboxylic acid

OR23361-100MG OR23361-1G OR23361-5G
77407-276EA 21.26 USD
77407-276 77407-278 77407-280
1,3-dimethyl-1H-thieno[2,3-c]pyrazole-5-carboxylic acid 95%
1,3-dimethyl-1H-thieno[2,3-c]pyrazole-5-carboxylic acid
Formula: C₈H₈N₂O₂S
MW: 196.224 g/mol
Melting Pt: 260 °C
Storage Temperature: Ambient
MDL Number: MFCD00126746
CAS Number: 25252-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR