2-Adamantylamine hydrochloride

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Aminoadamantane hydrochloride, Tricyclo[3.3.1.1(3,7)]dec-2-ylamine hydrochloride, 2-Adamantanamine hydrochloride, Adamantan-2-amine hydrochloride, 2-Aminotricyclo[3.3.1.1(3,7)]decane hydrochloride, 2-Adamantanammonium chloride

OR22317-1G OR22317-25G OR22317-5G
77406-868EA 24.46 USD
77406-868 77406-870 77406-872
2-Adamantylamine hydrochloride
2-Adamantylamine hydrochloride
Formula: C₁₀H₁₈ClN
MW: 187.71 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00074743
CAS Number: 10523-68-9
EINECS: 234-074-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR