4-Nitrophenylhydrazine hydrochloride

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-(4-Nitrophenyl)hydrazine hydrochloride, (4-Nitrophenyl)hydrazine hydrochloride, 4-Nitrophenylhydrazinium chloride

OR1938-100G OR1938-25G OR1938-5G
77406-448EA 158.94 USD
77406-448 77406-450 77406-452
4-Nitrophenylhydrazine hydrochloride
4-Nitrophenylhydrazine hydrochloride
Formula: C₆H₇N₃O₂·HCl
MW: 189.6 g/mol
Melting Pt: 198…200 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00040592
CAS Number: 636-99-7
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR