1,1'-Hexamethylenebis[5-(2-ethylhex-1-yl)]biguanide dihydrochloride

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Alexidine dihydrochloride, N,N''''-Hexane-1,6-diylbis[N'-(2-ethylhex-1-yl)(imidodicarbonimidic diamide)] dihydrochloride

OR2503-1G OR2503-250MG
77407-584EA 186.44 USD
77407-584 77407-586
1,1'-Hexamethylenebis[5-(2-ethylhex-1-yl)]biguanide dihydrochloride
1,1'-Hexamethylenebis[5-(2-ethylhex-1-yl)]biguanide dihydrochloride

Has a antibacterial, antiplaque properties against Streptococcus mutans, Actinomyces ciscosus, and Actinomyces naesludii.

Formula: C₂₆H₅₆N₁₀·2ClH
MW: 581.712 g/mol
Melting Pt: 220…224 °C
Storage Temperature: Freezer
MDL Number: MFCD00082369
CAS Number: 1715-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR