(4-(1H-1,2,4-Triazol-1-yl)phenyl)methanol
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
4-(1,2,4-Triazol-1-yl)benzyl Alcohol
OR23307-10G
OR23307-1G
OR23307-5G
77407-234EA
159.96
USD
77407-234
77407-236
77407-238
(4-(1H-1,2,4-Triazol-1-yl)phenyl)methanol
(4-(1H-1,2,4-Triazol-1-yl)phenyl)methanol
Formula:
C₉H₉N₃O MW: 175.1873 g/mol Melting Pt: 124…126 °C Storage Temperature: Ambient |
MDL Number:
MFCD02682061 CAS Number: 143426-50-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...