3-Benzo[b]thiophenesulfonyl chloride 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-Benzothiophene-3-sulphonyl Chloride, Benzothiophene-3-sulphonyl chloride

OR23173-1G OR23173-250MG
77407-156EA 124.3 USD
77407-156 77407-158
3-Benzo[b]thiophenesulfonyl chloride 95%
3-Benzo[b]thiophenesulphonyl chloride
Formula: C₈H₅ClO₂S₂
MW: 232.71 g/mol
Melting Pt: 85…88 °C
Storage Temperature: Ambient
MDL Number: MFCD02681897
CAS Number: 18494-87-6
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR