2-(Bromomethyl)benzothiazole

Supplier: APOLLO SCIENTIFIC
Danger

OR23090-1G OR23090-50MG OR23090-5G
77407-082EA 62.16 USD
77407-082 77407-084 77407-086
2-(Bromomethyl)benzothiazole
2-(Bromomethyl)benzothiazole
Formula: C₈H₆BrNS
Melting Pt: 47…49 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00461195
CAS Number: 106086-78-6
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR