2,2'-[ethylenebis(oxy)]bisacetic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3,6-Dioxaoctanedioic acid

OR19555-1G OR19555-5G
77406-492EA 41.78 USD
77406-492 77406-494
2,2'-[ethylenebis(oxy)]bisacetic acid
2,2'-[ethylenebis(oxy)]bisacetic acid
Formula: C₆H₁₀O₆
MW: 178.14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00054502
CAS Number: 23243-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR