3-Butyne-1-ol

Supplier: APOLLO SCIENTIFIC
Warning

OR1952-100G OR1952-25G OR1952-500G
77406-480EA 66.22 USD
77406-480 77406-482 77406-484
3-Butyne-1-ol
3-Butyne-1-ol
Formula: C₄H₆O
Boiling Pt: 128…130 °C
Melting Pt: –63.6 °C
Density: 0.926 g/cm³ (25 °C)
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00002955
CAS Number: 927-74-2
EINECS: 213-161-9
UN: 1986
ADR: 3+6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR