2-Methyl-1,3-benzoxazol-5-amine

Supplier: APOLLO SCIENTIFIC

Synonyms: 5-Amino-2-methyl-1,3-benzoxazole, 2-Methylbenzo[d]oxazol-5-amine

OR1878-1G OR1878-500MG
77406-398EA 171.16 USD
77406-398 77406-400
2-Methyl-1,3-benzoxazol-5-amine
2-Methyl-1,3-benzoxazol-5-amine
Formula: C₈H₈N₂O
MW: 148.161 g/mol
MDL Number: MFCD00628970
CAS Number: 72745-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR