1,1'-Carbonyldiimidazole 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR18334-1KG OR18334-250G OR18334-50G
77406-238EA 121.24 USD
77406-238 77406-240 77406-242
1,1'-Carbonyldiimidazole 98%
1,1'-Carbonyldiimidazole
Formula: C₇H₆N₄O
Melting Pt: 117…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00005286
CAS Number: 530-62-1
EINECS: 208-488-9
UN: 3263
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR