4-Bromo-2,6-dinitrotoluene

Supplier: APOLLO SCIENTIFIC
Warning

OR17614-1G OR17614-5G
77406-058EA 34.26 USD
77406-058 77406-060
4-Bromo-2,6-dinitrotoluene
4-Bromo-2,6-dinitrotoluene
Formula: C₇H₅BrN₂O₄
MW: 261.03 g/mol
Melting Pt: 86…89 °C
Storage Temperature: Ambient
MDL Number: MFCD07357280
CAS Number: 95192-64-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR