2-Bromo-4-methyl-6-nitroaniline

Supplier: APOLLO SCIENTIFIC
Warning

OR22152-25G OR22152-5G
77406-806EA 50.94 USD
77406-806 77406-808
2-Bromo-4-methyl-6-nitroaniline
2-Bromo-4-methyl-6-nitroaniline
Formula: C₇H₇BrN₂O₂
Melting Pt: 64…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00209452
CAS Number: 827-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR