3-Bromo-2-nitroaniline 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR2214-25G OR2214-5G
77406-798EA 409.94 USD
77406-798 77406-800 77406-802
3-Bromo-2-nitroaniline 98%
3-Bromo-2-nitroaniline
Formula: C₆H₅BrN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD08063248
CAS Number: 7138-15-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR