2-(2-Methoxyphenyl)-5-oxotetrahydrofuran-3-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR21836-1G OR21836-5G
77406-724EA 41.78 USD
77406-724 77406-726 77406-728
2-(2-Methoxyphenyl)-5-oxotetrahydrofuran-3-carboxylic acid
2-(2-Methoxyphenyl)-5-oxotetrahydrofuran-3-carboxylic acid
Formula: C₁₂H₁₂O₅
Melting Pt: 124…125 °C
Storage Temperature: Ambient
MDL Number: MFCD02677730
CAS Number: 117621-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR