ω,2,4-Trichloroacetophenone

Supplier: APOLLO SCIENTIFIC
Danger

OR18371-100G OR18371-500G
77406-306EA 21.34 USD
77406-306 77406-308
ω,2,4-Trichloroacetophenone
ω,2,4-Trichloroacetophenone
Formula: C₈H₅Cl₃O
Boiling Pt: 130…135 °C/4 mmHg
Melting Pt: 47…56 °C
Density: 1.312 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000934
CAS Number: 4252-78-2
EINECS: 224-218-2
UN: 2923
ADR: 8+6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR