2,4-Dichlorobenzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR18360-100G OR18360-1KG OR18360-500G
77406-276EA 21.26 USD
77406-276 77406-278 77406-280
2,4-Dichlorobenzoic acid
2,4-Dichlorobenzoic acid
Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
Melting Pt: 159…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00002414
CAS Number: 50-84-0
EINECS: 200-067-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR