N-Butylaniline 99%

Supplier: APOLLO SCIENTIFIC
Warning

OR18275-100G OR18275-500G OR18275-50G
77406-198EA 28.52 USD
77406-198 77406-200 77406-202
N-Butylaniline 99%
N-Butylaniline
Formula: (C₆H₅)NHCH₂CH₂CH₂CH₃
MW: 149.24 g/mol
Boiling Pt: 236…242 °C
Melting Pt: –14.4 °C
Density: 0.932 g/cm³ (25 °C)
Flash Pt: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD00035798
CAS Number: 1126-78-9
EINECS: 214-425-6
UN: 2738
ADR: 6.1 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR