2-Methyl-4-pyridineboronic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR2618-1G OR2618-5G
77407-744EA 21.4 USD
77407-744 77407-746
2-Methyl-4-pyridineboronic acid
2-Methyl-4-pyridineboronic acid
Formula: C₆H₈BNO₂
Melting Pt: 122…123 °C
Storage Temperature: Freezer
MDL Number: MFCD03095364
CAS Number: 579476-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR