1-Isopropenyl-4-methoxybenzene 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Methoxy-4-(1-propen-2-yl)benzene, 4-Isopropenylanisole

OR2606-1G OR2606-250MG OR2606-5G
77407-736EA 119.2 USD
77407-736 77407-738 77407-740
1-Isopropenyl-4-methoxybenzene 95%
1-Isopropenyl-4-methoxybenzene
Formula: C₁₀H₁₂O
MW: 148.21 g/mol
Boiling Pt: 130 °C/1.5 mmHg
Melting Pt: 30…32 °C
Density: 0.986 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD09414742
CAS Number: 1712-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR