3,5-Dioxocyclohexanecarboxylic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR2410-1G OR2410-25G OR2410-5G
77407-414EA 44.82 USD
77407-414 77407-416 77407-418
3,5-Dioxocyclohexanecarboxylic acid 97%
3,5-Dioxocyclohexanecarboxylic acid
Formula: C₇H₈O₄
Melting Pt: 180…184 °C
Storage Temperature: Ambient
MDL Number: MFCD09836162
CAS Number: 42858-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR