3-(Aminomethyl)-2,5-dimethylfuran 90%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (2,5-Dimethylfur-3-yl)methylamine

OR22999-1G OR22999-250MG
77407-006EA 406.5 USD
77407-006 77407-008
3-(Aminomethyl)-2,5-dimethylfuran 90%
3-(Aminomethyl)-2,5-dimethylfuran
Formula: C₇H₁₁NO
MW: 125.168 g/mol
Boiling Pt: 42…56 °C (0.1 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD02677701
CAS Number: 306934-85-0
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR