3',5'-Di-tert-butyl-4'-hydroxyacetophenone

Supplier: APOLLO SCIENTIFIC
Warning

OR21943-25G OR21943-5G
77406-748EA 458.46 USD
77406-748 77406-750
3',5'-Di-tert-butyl-4'-hydroxyacetophenone
3',5'-Di-tert-butyl-4'-hydroxyacetophenone
Formula: C₁₆H₂₄O₂
MW: 248.37
Melting Pt: 146…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00017520
CAS Number: 14035-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR