1,4-Dihydropyrazine-2,3-dione
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR21538-1G
OR21538-250MG
77406-706EA
61.12
USD
77406-706
77406-708
1,4-Dihydropyrazine-2,3-dione
1,4-Dihydropyrazine-2,3-dione
Formula:
C₄H₄N₂O₂ MW: 112.087 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00275454 CAS Number: 931-18-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...