(2-Bromo-5-chlorophenyl)acetic acid
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR2153-100G
OR2153-10G
OR2153-25G
77406-698EA
516.54
USD
77406-698
77406-700
77406-702
(2-Bromo-5-chlorophenyl)acetic acid
(2-Bromo-5-chlorophenyl)acetic acid
Formula:
C₈H₆BrClO₂ Melting Pt: 101…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153890 CAS Number: 81682-38-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...