3-Aminopiperidine-2,6-dione hydrochloride 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR20048-100G
OR20048-25G
OR20048-500G
77406-546EA
166.06
USD
77406-546
77406-548
77406-550
3-Aminopiperidine-2,6-dione hydrochloride 97%
3-Aminopiperidine-2,6-dione hydrochloride
Formula:
C₅H₉ClN₂O₂ Melting Pt: 245 °C Storage Temperature: Ambient |
MDL Number:
MFCD11042437 CAS Number: 24666-56-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...