Ethyl-3,4-diaminobenzoate

Supplier: APOLLO SCIENTIFIC
Warning

OR2001-100G OR2001-25G OR2001-5G
77406-536EA 111.04 USD
77406-536 77406-538 77406-540
Ethyl-3,4-diaminobenzoate
Ethyl-3,4-diaminobenzoate
Formula: C₉H₁₂N₂O₂
Melting Pt: 108…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00179337
CAS Number: 37466-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR