2-(4-Hydroxyphenyl)propionic acid

Supplier: APOLLO SCIENTIFIC
Danger

OR17909-10G OR17909-1G OR17909-50G
77406-150EA 115.12 USD
77406-150 77406-152 77406-154
2-(4-Hydroxyphenyl)propionic acid
2-(4-Hydroxyphenyl)propionic acid
Formula: C₉H₁₀O₃
Melting Pt: 130…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00128847
CAS Number: 938-96-5
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR